Sodium diphenylamine-4-sulphonate 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium (4-anilinobenzenesulphonate), Diphenylamine-4-sulphonic acid, sodium salt
A19768.06 A19768.14
A19768.06EA 65.1 EUR
A19768.06 A19768.14
Sodium diphenylamine-4-sulphonate 98%
Sodium diphenylamine-4-sulphonate
Purity: 98% CAS No.: 6152-67-6 Molecular Formula: C12H10NNaO3S Formula Weight: 271.27 MDL No.: MFCD00036468
Formula: C₁₂H₁₀NNaO₃S
MW: 271.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397