Triacetoxyethylsilane 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethylsilanetriyl triacetate, Triacetoxy(ethyl)silane

B23187.22 B23187.36
B23187.22EA 38.3 EUR
B23187.22 B23187.36
Triacetoxyethylsilane 96%
Triacetoxyethylsilane

Purity: 96% CAS No.: 17689-77-9 Molecular Formula: (CH3CO2)3SiCH2CH3 Formula Weight: 234.28 Flash Point: 106°(224°F) Refractive Index: 1.4130 Melting Point: 7-9° Boiling Point: 97-99°/4mm UN No.: 1760 MDL No.: MFCD00053703

Formula: C₈H₁₄O₆Si
Boiling Pt: 97…99 °C (4 mmHg)
Melting Pt: 7…9 °C
Density: 1.140
Flash Pt: 106 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053703
CAS Number: 17689-77-9
EINECS: 241-677-4
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR