1,2,3-Triazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
333662-1G 333662-5G
SIAL333662-1GEA 54 EUR
SIAL333662-1G SIAL333662-5G
1,2,3-Triazole, Sigma-Aldrich®
1,2,3-Triazole
Formula: C₂H₃N₃
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 23…25 °C
Density: 1.192 g/cm³ (20 °C)
MDL Number: MFCD00014490
CAS Number: 288-36-8