Sterilisation Bags, Heat Seal
Supplier: S▄DPACK MEDICA
7600516 7600536 7600507 7600524 7600530 7600525
300-0233EA 24.1 EUR
300-0233 300-0234 129-1404 129-1427 129-1407 129-1438
Sterilisation Bags, Heat Seal
Bags Sterilisation Pouches
Heat-sealed bags for sterilisation with paper on one side (60 g/m²) and assembled, transparent PET/PP plastic on the other side.
  • Dual sterilisation displays for autoclave and ethylene oxide
  • Welded joint
  • Available in different sizes