Bags, biohazard, B-Safe
Supplier: Alpha Packaging
121-072 121-073 121-027 121-028 121-029
129-0351EA 54.3 EUR
129-0351 129-0352 129-0339 129-0340 129-0341
Bags, biohazard, B-Safe
Bags Waste Disposal Bags
Biohazard bags in PE, 125 µm (500 gauge), yellow.
  • For heavy duty disposal
  • Printed with biohazard symbol and the wording `Biohazard, Danger of Infection' and `Burn Without Opening' on the two largest bags
  • Warning in English, French and German