VWR®, Beakers, Low Form
Supplier: VWR Collection
213-1621EA 25 EUR
213-1621 213-1622 213-1623 213-1624 213-1642 216-1643 219-1644 222-1645 213-1725 213-1726
VWR®, Beakers, Low Form
Beakers
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use at +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
ISO 7056
Certifications: ISO 7056