Beakers, low form, Nalgene®
Supplier: Thermo Fisher Scientific
1203-0030 1203-0050 1203-0100 1203-0150 1203-0250 1203-0400 1203-0600 1203-1000 1203-2000 1203-4000
213-7130EA 0 EUR
213-7130 213-7131 213-7132 213-7133 213-7134 213-7135 213-7136 213-7137 213-7138 213-7139
Beakers, low form, Nalgene®
Beakers
PMP (TPX) crystal clear.
  • Good chemical resistance
  • Heat resistant up to +150 °C, autoclavable at +121 °C (max. 20 minutes)
  • Smooth surfaces for easy, fast cleaning
Designed for general laboratory use. With spout and easy to read, printed graduations with size code, plastic code, maximum temperature for use and 'Not flameproof' symbol. Ribbed for easy stacking. No drip, single spout design provides precision pouring control.
ISO 7056
Certifications: ISO 7056