VWR®, Beakers, Tall Form
Supplier: VWR Collection
213-1169EA 61.1 EUR
213-1169 213-1170 213-1171 213-1172 213-1173 213-1174 213-1175 213-1176 213-1178 213-0470
VWR®, Beakers, Tall Form
Beakers
Borosilicate 3.3 glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
  • Volume graduations and labelling area
The economical alternative for a wide range of laboratory applications.
DIN 12331, ISO 3819
Certifications: DIN 12331, ISO 3819