Carboys, narrow neck, LDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
2220-0010 2220-0020
216-4398EA 37 EUR
216-4398 216-4399
Carboys, narrow neck, LDPE, with screw caps, Nalgene®
Bottles Carboys
LDPE, translucent with white PP screw cap.
  • Good chemical resistance
  • Temperature resistant from −100 to +80 °C
  • Leakproof
Large handle makes carrying easier while wearing rubber gloves for safe handling of corrosive liquids. Opening offset to one side for easy, precise pouring.