Dropping bottles, Lucas
Supplier: MUELLER KREMPEL
14.196.03 14.196.04 14.196.05 14.196.07 14.196.01 14.196.06 14.196.02
215-9102EA 110 EUR
215-9102 215-9103 215-9104 215-9106 MUEL14.196.01 215-3583 MUEL14.196.02
Dropping bottles, Lucas
Bottles Dropping Bottles
Amber glass, without closure.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Please order screw caps and drop dispensers separately
Brown coloured to protect light-sensitive materials.