Culture media bottles, square, with septum caps, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
342023-0060 342023-0125 342023-0500 342023-1000 342023-9500
215-1165EA 1420 EUR
215-1165 215-1489 215-1490 215-1491 215-1492
Culture media bottles, square, with septum caps, sterile, Nalgene®
Bottles Cell Culture Bottles
PETG, clear with silicone/PTFE screw cap.
  • Good chemical resistance
  • Temperature resistant from −40 to +70 °C
  • The silicone/PTFE cap with septum is provided with an inert and repellent PTFE finish to ensure the integrity of samples
Ideal for packaging biological and diagnostic reagents, for which aseptic removal is necessary or to which supplements are to be added using a syringe. Bottle and caps are gamma-sterilised and non-toxic.
Certifications: Complying with the requirements of USP, Class VI and European Pharmacopeia for Abnormal Toxicity Standards.
Packaging: Packed in shrink-wrapped layers. Overwrapped in double-packed boxes for clean room applications.