VWR®, Bottles, Narrow Neck, Round, LDPE, with Screw Caps
Supplier: VWR Collection
215-5637EA 12.7 EUR
215-5637 215-5638 215-5639 215-5640 215-5641 215-5642
VWR®, Bottles, Narrow Neck, Round, LDPE, with Screw Caps
Bottles Plastic Bottles
LDPE, translucent. Excellent for sample collection, storage and transporting liquids.
  • Leakproof, robust and versatile
  • Squeezable, containers can be used as a dropper or wash bottle
  • Raised graduations
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904