Laboratory bottles, narrow neck, with thread, amber glass, DURAN®
Supplier: DWK Life Sciences
218061409 218061709 218062405 218063607 218064406 218065402 218066304 218067309 218068605 218068802 218069104 218062902 218065102 218066904 218061458 218061758 218062454 218062951 218063656 218064455 218065151 218065451 218066353 218066953 218067358 218068654 218060807 218060856
215-1572EA 35.2 EUR
215-1572 215-1573 215-1574 215-1575 215-1576 215-1577 215-1578 215-1579 215-1580 215-1581 215-1582 215-2434 215-2435 215-2436 215-4487 215-3793 215-3794 215-3796 215-3797 215-3798 215-3804 215-3805 215-3799 215-3806 215-3807 215-3808 215-3296 215-3215
Laboratory bottles, narrow neck, with thread, amber glass, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, amber, without or with pouring ring and screw cap.
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Purely external application of the colour ensures unchanged DURAN® properties on the interior
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
  • 10 ml bottles do not conform with ISO 4796-1 standard; specially shaped glass edge for improved pouring out (so that an additional plastic pouring ring is not required)
  • 20 ml bottles are made with specially shaped glass edge for improved pouring out (so that an additional plastic pouring ring is not required)
Round, with graduation and DIN thread.
With printed Retrace Code (batch identification).
With specially shaped glass rim for improved pouring; no additional pouring ring is needed.
Ring-shaped reinforcement on the bottle shoulder makes the filling line of the nominal volume visible.
Certifications: ISO 4796-1