Bottles, narrow neck, round, PFA, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
1630-0004 1630-0008 1630-0016 1630-0032 DS1630-0001 DS1630-0002
215-7163EA 129 EUR
215-7163 215-7164 215-7165 215-7166 215-7161 215-7162
Bottles, narrow neck, round, PFA, with screw caps, Nalgene®
Bottles Plastic Bottles
PFA, translucent, with PFA screw cap.
  • Highest chemical resistance
  • Heat resistant from −270 to +250 °C, autoclavable
  • Leakproof closure
Bottles with the highest resistance to  extreme temperatures, chemicals and corrosion. Inert to practically all chemicals with the exception of molten alkaline metals, fluorine compounds and complex halogen compounds at high temperatures and under pressure.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.