Bottles, narrow neck, rectangular, LDPE, with thread
Supplier: MUELLER KREMPEL
14.258.01 14.258.02 14.258.03 14.258.04
215-8331EA 2.1 EUR
215-8331 215-8332 215-8333 215-8334
Bottles, narrow neck, rectangular, LDPE, with thread
Bottles Plastic Bottles
LDPE, translucent, without screw cap. Ideal for storing and shipping liquids. Graduated.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Side ribbing for better grip
Ordering information: The bottles are supplied without caps. For suitable caps for 125/500 and 1000 ml please see 215-9226 (for 250 ml botlles see MUEL14.063.01).