Bottle, narrow neck, round, PP, with screw caps
Supplier: KARTELL
1595 1597 1599 1601 1603
215-1334EA 4.6 EUR
215-1334 215-1335 215-1336 215-1337 215-1338
Bottle, narrow neck, round, PP, with screw caps
Bottles Plastic Bottles
In PP, with threaded cap.
  • With standardised GL thread
  • Perfect sealing
Certifications: Compliant with DIN 13316 and DIN 168.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904