VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
215-5630EA 13.6 EUR
215-5630 215-5631 215-5632 215-5633 215-5634 215-5635 215-5636
VWR®, Bottles, Wide Neck, with Screw Cap
Bottles Plastic Bottles
LDPE, translucent.
  • Good chemical resistance
  • Leakproof, tough and versatile
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The wide mouth allows easy filling and cleaning. Without graduations.