Bottles, wide neck, with screw cap, PassPort IP 2, Nalgene®
Supplier: Thermo Fisher Scientific
2199-0004 2199-0008 2199-0016 2199-0032
215-1043EA 44.6 EUR
215-1043 215-1044 215-1045 215-1046
Bottles, wide neck, with screw cap, PassPort IP 2, Nalgene®
Bottles Plastic Bottles
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 120 °C
  • Leakproof closure
Safe, durable, economical bottles for the transport and packaging of hazardous goods. Recommended for customers who wish to put together and certify their own packaging system. Extra-wide mouth for simple filling and emptying of liquids and dry substances. The sturdy walls withstand cracking and damage from impact caused by shocks during transport. The bottom of the bottle incorporates IP2 marking, material, volume and registration notch. Evaluated at 15 psi per 49 CFR 173.27 © (2), ICAO Technical Instructions Part 4; 1.1.6 and IATA Dangerous Goods Regulations, Section 5.0.2.9.