Bottles, wide neck, clear, with GLS 80® thread, DURAN®
Supplier: DWK Life Sciences
1112627 1112713 1112715 1113949 1113950 1113951 1200154 218606953 218603656 1200265
215-1785EA 29.3 EUR
215-1785 215-1786 215-1787 215-1789 215-1790 215-1791 215-4177 215-2850 215-2955 215-4178
Bottles, wide neck, clear, with GLS 80® thread, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear, with screw caps and pouring rings.
  • Excellent chemical resistance, high temperature resistance
  • Extra wide opening allows easy cleaning
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Round, with graduations and specially developed GLS 80® thread. Opening and closing the bottle is easy with quick closure mechanism. The 80 mm wide outer diameter of the bottle neck enables bottles to be filled conveniently and safely with powders, pastes,More Product Information