Bottles, wide neck, with screw cap, Corning® Gosselin™
Supplier: Corning
LR1000-01 LR1000-03 LR1000-04 LR1000-05 LR1000-08 LR1000-09 LR100-01 LR100-03 LR100-04 LR100-05 LR100-12 LR150A-01 LR150A-02 LR150A-03 LR150A-07 LR2000-01 LR2000-03 LR250-01 LR250-02 LR250-04 LR250-05 LR250-07 LR250-09 LR250-11 LR500-01 LR500-02 LR500-04 LR500-05 LR500-06 LR500-07 LR100-10 LR100-07 LR100-08 LR100-13 LR1000-17 LR1000-19 LR150A-06 LR150A-08 LR250-14 LR250-16 LR500-15 LR500-20 LR50A-02 LR50A-05 LR50A-01 LR50A-12 LR50A-18 LR1000-02 LR150A-04 LR250-06 LR500-03
215-3962EA 160 EUR
215-3962 215-0744 215-0743 215-0742 215-0741 215-3965 215-3290 215-0749 215-0748 215-0747 215-0745 215-0752 215-0751 215-0754 215-0753 215-0758 215-0759 215-1295 215-0765 215-0764 215-3970 215-0762 215-0761 215-3971 215-0769 215-0768 215-0767 215-0771 215-0770 GOSSLR500-07 GOSSLR100-10 GOSSLR100-07 GOSSLR100-08 GOSSLR100-13 215-3968 215-3969 GOSSLR150A-06 GOSSLR150A-08 215-3972 215-3973 GOSSLR500-15 215-3977 215-3238 215-2183 215-2184 215-3239 215-3240 215-0740 215-0755 215-0760 215-0766
Bottles, wide neck, with screw cap, Corning® Gosselin™
Bottles Plastic Bottles
HDPE, translucent with screw caps.
  • Bottles are supplied capped aseptic or ionised
  • Excellent shock resistance
  • Wide neck, easy to fill
A range of round wide mouth bottles. Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.