Woulff bottles, DURAN®
Supplier: DWK Life Sciences
247094403 247095408 247096301 247097306
189-1101EA 129 EUR
189-1101 189-1102 189-1103 189-1104
Woulff bottles, DURAN®
Bottles Woulff Bottles
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
With 3 standard ground joint necks. Vacuum tight.
Certifications: DIN 12480.