Transport and Storage Boxes
Supplier: LINPAC
3127-NA 1201 1100 1102 1104 1103 1108
216-1761EA 28.1 EUR
216-1761 216-1762 216-1763 216-1764 216-1765 216-1767 216-1768
Transport and Storage Boxes
Boxes Storage Boxes
Plastic, white.
  • With tightly fitting lid
  • Suitable for use at −50 °C
Storage boxes made of polystyrene, white and lid made of soft polyethylene.