Accessories for VWR® Protecting Covers for Clamp jaws
Supplier: VWR Collection
241-0149EA 25.3 EUR
241-0149 241-0150 241-0151
Accessories for VWR® Protecting Covers for Clamp jaws
Clamps General Purpose Clamps
These protecting covers for clamp jaws are made from red vinyl.
  • Good chemical and thermal stability
  • Fits snugly but easily over the clamp jaws
  • Protects clamped items
Suitable for VWR® clamps with rounded jaws.