Clamps
Supplier: BOCHEM
5404 5450 5451 5403 5401 5402 5452
241-0400EA 47.5 EUR
241-0400 241-0401 241-0402 241-1112 241-1110 241-1111 241-0403
Clamps
Clamps General Purpose Clamps
Stainless steel.
  • Jaws are covered with a layer of cork to protect clamped items
  • Without bosshead for fixing to stand
  • Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894