Clamps
Supplier: BOCHEM
5461 5462 5463
241-7188EA 18.1 EUR
241-7188 241-7189 241-7190
Clamps
Clamps General Purpose Clamps
Malleable cast iron, chromed.
  • Jaws have cork inserts to protect clamped items
  • Without bosshead for fixing to stand
  • Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894