Pouring rings for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
292421907 292422809 292441909 292442802
217-0202EA 5.8 EUR
217-0202 217-0203 217-0204 217-0205
Pouring rings for laboratory bottles, DURAN®
Closures Bottle Closures
PP, blue or ETFE, red.
  • For DIN GL threads
  • PP: heat resistant up to +140 °C, autoclavable
  • ETFE: heat resistant up to +180 °C, autoclavable and sterilisable with hot air