Screw caps, tamper evident, for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
1017526 1155886
215-1101EA 22.7 EUR
215-1101 215-2445
Screw caps, tamper evident, for laboratory bottles, DURAN®
Closures Bottle Closures
PP.
  • Very good chemical resistance
  • Heat resistant up to +140 °C
  • High leak tightness
For laboratory bottles with DIN thread GL 45.