Screw cap for laboratory bottles, GLS 80®, quick release, DURAN®
Supplier: DWK Life Sciences
1112716 1160166 1165888 1167307 1152921
215-1788EA 34.4 EUR
215-1788 215-2085 215-2292 215-2293 215-3999
Screw cap for laboratory bottles, GLS 80®, quick release, DURAN®
Closures Bottle Closures
PP, blue.
With lip seal
Heat resistant to 140 °C, autoclavable
Easy to use, special thread design requires less than two turns to open or close
A matching PP (blue) pouring ring is also available, permitting clean, drop-free use
PSU compound, white
With PTFE coated seal. The material used is a special compound based on polyacrylsulphone. Consequently the chemical, thermal and mechanical properties of the material are noticeably improved and matched to laboratory requirements. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly closed.
Temperature resistant to 180 °C, autoclavable
GLS 80® bottle can be opened and closed with only a three-quarter turn
A matching PTFE pouring ring is also available, permitting clean, drop-free use
With PTFE coated seal. The material used is a special compound based on polyacrylsulphone. Consequently the chemical, thermal and mechanical properties of the material are noticeably improved and matched to laboratory requirements. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly closed.