Laboratory and chemical protection apron, PVC
Supplier: Elbe-Werkstätten GmbH
090-046 090-047
113-3317EA 44.4 EUR
113-3317 113-3318
Laboratory and chemical protection apron, PVC
Workwear
The transparent PVC apron is resistant to most organic acids and alkalis. The halter neck and side ties are made from strong polyester fabric.
  • With high-frequency welded eyelets
  • Easy to clean: machine washable at 60 °C
  • Also suitable for use in the food and health industries
Thickness: 0,3 mm
Colour: natural