Overalls, KLEENGUARD* A40
Supplier: KIMBERLY CLARK
9794 9793 9792 9795 9790 9791
113-7936EA 315 EUR
113-7936 113-7935 113-7934 113-7937 113-7945 113-7933
Overalls, KLEENGUARD* A40
Disposable Garments
This hooded overall with full length zip is made from ultra-strong, silicone-free, anti-static fabric. It provides protection against chemical splashes and particles.
  • Laminate film provides a barrier against a wide range of chemicals (Type 6)
  • Particle protection, keeps out >99% of fibres larger than 1 µm (Type 5)
  • Strong triple stitched seams help protect against tearing
  • Hood is designed to allow a mask to be worn and preserves freedom of movement
  • Ultra-low lint performance
Especially well suited for critical production, such as pharmaceutical, chemical handling, manufacturing and electronic assembly.
Colour: White
Certifications: Type 5/6, EN 1149-1, EN 1073-2; Cat. III