Distillation columns, Vigreux
Supplier: Lenz Laborglas GmbH & CO.KG
05130201 05130303 05130304 05130503 05130504 05130301
201-3216EA 59.7 EUR
201-3216 201-0351 201-3215 201-1001 201-1173 201-1000
Distillation columns, Vigreux
Distillation Components Distillation Columns
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Air cooled fractionating column with alternating horizontal and inclined indentations.