Capillary columns, BP5
Supplier: Trajan Scientific and Medical
054167 054168 054177 054178 054180 054182 054183 054184 054186 054195 054199 054188 054189 054196 054203 054216
558-0141EA 404 EUR
558-0141 558-0142 568-0132 558-0152 568-0134 558-0144 558-0145 558-0146 558-0154 558-0160 568-0140 568-0135 568-0136 568-0137 568-0141 568-0144
Capillary columns, BP5
Chromatography Columns
A general purpose phase with excellent high temperature characteristics. Popular column used for a wide variety of applications.
5% phenyl/95% dimethyl polysiloxane.
Max. temperature: 340 °C.
Suitable replacement for: CP-Sil 8CB, DB-5, HP-5, MDN-5, PTE-5, Rtx-5, SPB-5, Ultra-2, VB-5, ZB-5.