Reversed phase HPLC columns, HxSil C8 and C18
Supplier: HAMILTON BONADUZ
79868 79135 79137 79138 79107 79108 79109 79116 79117 79882 79889 79451 79452 79458 79459 79869 79299 79923 79922
554-1331EA 800 EUR
554-1331 554-1235 554-1237 554-1238 554-1207 554-1208 554-1209 554-1216 554-1217 554-1340 554-1347 554-1266 554-1267 554-1272 554-1273 554-1332 HAMI79299 HAMI79923 HAMI79922
Reversed phase HPLC columns, HxSil C8 and C18
Chromatography Columns
Silica-based 100 Å columns for general reversed phase separations.
  • Separation of closely related PAHs
  • Better retention of poorly retained compounds
  • Separation of neutral, acidic and basic compounds without tailing (tailing factor of less than 2,0)