Allihn condensers with standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
05223302 05223303 05223304 05223403 05223404 05223504 05223201 05223202 05223204 05223301 05223203
201-1687EA 71.6 EUR
201-1687 201-1688 201-0360 201-1689 201-3341 LENZ05223504 201-1686 201-1685 LENZ05223204 LENZ05223301 LENZ05223203
Allihn condensers with standard ground joints
Condensers
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With five 'balls' (up to 250 mm jacket length) or eight 'balls' (for 400 mm jacket length), as well as standard ground joint cone and sleeve. Water connection via glass olive.
Compliant with DIN 12581 or supplementary DIN
Certifications: Compliant with DIN 12581 or supplementary DIN