Screwthread couplings for threaded pipes, DURAN®
Supplier: DWK Life Sciences
292260556 292260659 292260856 292260959 292261055
201-1705EA 22.6 EUR
201-1705 201-1706 201-1708 201-1707 201-0493
Screwthread couplings for threaded pipes, DURAN®
Fittings and Connectors
PBT, red with integrated silicone (VMQ) PTFE seal.
  • For flexible connection of two DIN glass threads
  • Heat resistant from -45 to +180 °C
  • Extremely leak tight