Seals for threaded connection caps (Septa)
Supplier: BIRZLE PETER
292460503 292460606 292460906 1095576
201-1961EA 5.5 EUR
201-1961 201-1962 201-1963 201-1965
Seals for threaded connection caps (Septa)
Septa
Silicone, transparent.
  • Ideal for use as septa (for puncturing)
  • Very good chemical resistance
  • Very good thermal resistance
For threaded connection caps with GL thread.