Desiccators, DURAN®
Supplier: DWK Life Sciences
247814603 247815702 247816107 247816604 247816904 247824604 247825703 247826108 247826605 247826905 247826954 247825752 247826157 247826654
467-0055EA 169 EUR
467-0055 467-1134 467-1135 467-0056 467-0057 467-1137 467-1133 467-1136 467-0058 467-1132 SCOT247826954 SCOT247825752 SCOT247826157 SCOT247826654
Desiccators, DURAN®
Desiccators Desiccators, Glass
Clear DURAN®, borosilicate glass 3.3, choice of models with flat flange, NOVUS NS 24/29 junction, and outlet in lid with stopcock or desiccators with flat flange without junction and lid with a knob.
  • Ideal for vacuum and non vacuum applications
  • Supplied with lid
Certifications: According to DIN ISO 13130
Ordering information: Most are supplied without a plate, please order suitable plate separately.