Desiccators, MOBILEX type, DURAN®
Supplier: DWK Life Sciences
247855706 247856102 247856608 247856908
SCOT247855706EA 332 EUR
SCOT247855706 SCOT247856102 467-0059 467-0060
Desiccators, MOBILEX type, DURAN®
Desiccators Desiccators, Glass
DURAN® clear borosilicate glass 3.3, lid with threaded outlet, type MOBILEX (GL 32), stopcock with PTFE spindle.
  • Ideal for vacuum and non vacuum applications
  • Supplied with lid
Ordering information: Supplied without plate, please order suitable plate separately.