Desiccator bases, DURAN®
Supplier: DWK Life Sciences
247704604 247705703 247706108 247706605 247706905 247714605 247715704 247716109 247716606 247716906 247725705 247726101 247726607 247726907
467-1110EA 84.9 EUR
467-1110 467-1115 467-1120 467-1125 467-1130 467-1210 467-1215 467-1220 467-1225 467-1230 467-1315 467-1320 467-1325 467-1330
Desiccator bases, DURAN®
Desiccators Desiccator Bases
DURAN®, clear borosilicate glass 3.3. Ideal for drying and drying processes, storage under vacuum and protection from the effects of air and moisture. Applications include cultivation of organisms, material research and corrosion testing, cleaning equipment for analysis of trace elements, gas and liquid reactions and they can be used as a mini cleanroom for chip technology.
  • Choice of various desiccator components to suit different applications
  • Increased safety due to DURAN® quality borosilicate glass
  • All models have a flat flange, except 467-0139 which has a ring-grooved flange
  • Excellent mechanical, thermal and chemical resistance
Certifications: All model according to DIN ISO 13130, desiccators with a ring-grooved flange no DIN ISO size.
Ordering information: Supplied without plate, lid or stopcock, please order required accessories separately.