Desiccator plates
Supplier: MORGAN ADVANCED MATERIALS HALD
L119C/140 L119C/190 L119C/235 L119C/280 L119C/90
467-1915EA 52.8 EUR
467-1915 467-1920 467-1925 467-1930 467-1910
Desiccator plates
Desiccators Desiccators, Plates
Porcelain desiccator plates. Central hole 20 mm Ø, filter holes ~5 mm Ø. Suitable for use with any desiccators with corresponding diameter.
Certifications: Conform to DIN 12911