Cell culture dishes, Corning® Primaria™
Supplier: Corning
353801 353802 353803
734-0070EA 157.47 EUR
734-0070 734-0071 734-0072
Cell culture dishes, Corning® Primaria™
Culture Dishes Cell Culture Dishes
PS, sterile, with lid.
  • Unique, nitrogen-containing tissue culture surface chemistry improves attachment, spreading and growth for many primary cells or cell lines
  • Optically clear and no special storage required
  • Convenient, peel open, medical-style packaging
The gases used to manufacture Corning® Primaria™ contain both oxygen and ammonia, resulting in the incorporation in the surface of a variety of nitrogen-containing functional groups in addition to the negatively charged oxygen containing groups found on traditional tissue culture treated surfaces.More Product Information