VWR®, Dishes, Crystallising
Supplier: VWR Collection
216-0076EA 70.8 EUR
216-0076 216-0077 216-0060 216-0061 216-0062 216-0063 216-0064 216-0065 216-0066 216-0067 216-0068 216-0069 216-0070 216-0071 216-0072 216-0073 216-0074 216-0075 216-0078 216-0079
VWR®, Dishes, Crystallising
Trays/Dishes/Pots Crystallising Dishes
Borosilicate 3.3 glass, with or without spout.
  • Very good chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of applications. With white labelling area.
DIN 12337 (without spout)
DIN 12338 (with spout)