Petri dishes, Star™ Dish
Supplier: PHOENIX BIOMEDICAL
057 058 331 332 357 360 371 372 352 356 370 390 385 321 307 180 181 182 183 184 185 305 161 163 162 164 165 170 171 172 173 174 175 387 004
710-3512EA 157 EUR
710-3512 710-3513 710-3518 710-3519 710-3504 710-3505 710-3508 710-3509 710-3502 710-3503 710-3507 710-3511 710-3510 710-3516 391-0053 710-3521 710-0599 710-0600 710-0601 710-0602 710-0603 391-0531 391-0436 391-1003 391-1002 391-0421 391-0422 391-0437 391-0438 391-1004 391-1005 391-0423 391-0424 391-0425 391-0573
Petri dishes, Star™ Dish
Culture Dishes Petri Dishes
Clear PS. Ideal for colony counting.
  • Environmentally friendly manufacturing process uses reduced amounts of plastic
  • Sterile and particulate-free
90×15 mm dishes
The recessed lid on the nesting and semi-nesting ALPINE dish offers excellent stacking stability. It works with most models of media pouring equipment. The recess on the lid has a smoother bevelled angle allowing the dish to slide easily during automated pouring.
- Vents on the underside of the lid prevent condensation from developing and dripping onto the culture
- Extremely flat bottom allows aMore Product Information