Staining jars
Supplier: GLASWARENFABRIK KARL HECHT
42480010 42481010 42482010
631-9328EA 25.8 EUR
631-9328 631-9321 631-9329
Staining jars
Jars Staining Jars
Soda-lime glass.
  • For 10 standard slides  (76×26 mm)
Staining system contains staining jar, rack and handle made from stainless steel.
Ordering information: Insert and handle must be ordered separately.