Multiple use earplugs, 1261, Howard Leight®
Supplier: 3M
1261 1271
111-0039EA 168 EUR
111-0039 111-0040
Multiple use earplugs, 1261, Howard Leight®
Earplugs Reusable Earplugs
Earplugs manufactured from a soft elastic material to ensure a secure and comfortable fit.
  • Plugs can be inserted without squashing
  • Can be washed and reused many times
  • Unique design for easier insertion
Suitable for dirty or dusty environments.
SNR: 25 dB
EN 352-2
Certifications: EN 352-2
Delivery information: Supplied with a pocket case to protect the earplugs and keep them clean.
Caution: * One size fits all.