Bottle top vacuum filtration systems, without membrane, Nalgene®
Supplier: Thermo Fisher Scientific
300-4000 300-4050 300-4100
513-3001EA 244 EUR
513-3001 513-3002 513-3003
Bottle top vacuum filtration systems, without membrane, Nalgene®
Filters Bottle Top Filters
PS, reusable bottle top vacuum filtration systems. Suitable for analytical or sterile filtration, under vacuum or pressure.
  • Graduated receiver
  • Two side-arms for tubing 6 to 8 mm inner diameter
  • With cover for convenient sterile storage of filtrate
Compatible with 47/50 mm membranes.
Delivery information: Supplied with holder and receiver, analytical plate, sterilisation plate and cover.