Filter discs, VitraPOR®
Supplier: ROBU
VitraPOR®
511-0015EA 36.5 EUR
511-0015 511-0012 511-0496 511-0495 511-0011 511-0014 511-0013 511-0497 511-0492 511-0491 511-0494 511-0010 511-0493 511-0490 ROBU1530F ROBU5140F ROBU15309 ROBU15305 511-0389 511-0023 511-0386 511-0022 511-0388 511-0387 511-0024 511-0021 511-0020 511-0478 511-0477 511-0114 511-0479 511-0111 511-0474 511-0110 511-0473 511-0476 511-0113 511-0475 511-0112 511-0470 511-0472 511-0471 ROBU1520F ROBU5150F ROBU15205 ROBU15209 511-0247 511-0489 511-0488 511-0485 511-0484 511-0487 511-0486 511-0481 511-0480 511-0483 511-0482 511-0456 511-0455 511-0458 511-0457 511-0452 511-0451 511-0454 511-0453 511-0450 ROBU1550F ROBU15509 511-0449 ROBU15505 511-0448 511-0104 511-0467 511-0346 511-0103 511-0466 511-0345 511-0469 511-0348 511-0347 511-0468 511-0463 511-0100 511-0342 511-0341 511-0462 511-0102 511-0344 511-0465 511-0101 511-0343 511-0464 511-0461 511-0460 ROBU5160F 511-0459 511-0434 511-0433 511-0436 511-0435 ROBU15652 511-0430 ROBU15651 511-0432 ROBU15650 511-0310 511-0431 ROBU1540F ROBU15409 511-0309 511-0306 511-0427 511-0426 511-0429 511-0308 511-0307 ROBU15405 511-0428 511-0445 511-0444 511-0447 511-0446 511-0441 511-0440 511-0443 511-0442 ROBU5200F ROBU1565F ROBU5170F ROBU15655 511-0438 511-0437 511-0439 ROBU1570F 511-0410 511-0092 511-0091 511-0094 511-0093 511-0090 ROBU15705 ROBU15709 511-0409 511-0408 511-0405 511-0407 511-0406 511-0425 511-0420 ROBU5180F 511-0419 511-0416 511-0415 511-0418 511-0417 ROBU15053 ROBU15054 ROBU15051 ROBU1560F ROBU15052 511-0078 ROBU15055 511-0079 511-0074 511-0073 511-0070 511-0072 511-0071 ROBU15609 ROBU15605 511-0400 511-0084 511-0081 511-0080 511-0083 511-0082 ROBU5190F ROBU1505F ROBU1590F 511-0052 ROBU1510F 511-0051 511-0054 511-0053 511-0050 ROBU15105 ROBU15109 511-0063 ROBU1511F 511-0062 511-0064 511-0061 511-0060 ROBU1580F 511-0034 ROBU15255 511-0397 511-0396 511-0033 511-0399 511-0398 511-0393 511-0030 511-0392 511-0032 511-0395 ROBU1512F 511-0394 511-0031 ROBU5130F 511-0391 511-0390 511-0044 511-0041 511-0040 511-0043 511-0042 ROBU1525F ROBU15259
Filter discs, VitraPOR®
Filters Filter Discs
Filter discs made from DURAN®, borosilicate glass. Edges not fused and centred, surface untreated.
Porosity 0 (nominal pore size 160 - 250 µm): Gas distribution in liquids at low pressure, coarse filtration
Porosity 1 (nominal pore size 100 - 160 µm): Coarse filtration, gas distribution, liquid distribution, gas cleaning
Porosity 2 (nominal pore size 40 - 100 µm): Preparative fine filtration, mercury filtration, preparative work with coarse crystalline products
Porosity 3 (nominal pore size 16 - 40 µm): Analytical work with medium fine precipitates, fine gas filtration, analytical filtration, special filtration
Porosity 4 (nominal pore size 10 - 16 µm): Analytical fine filtration, analytical and preparative work with the finest precipitates, return and stop valve for mercury