Bottle top vacuum filtration systems, Nalgene®
Supplier: Thermo Fisher Scientific
DS0320-2545 DS0320-5033 DS0320-5045
516-0577EA 230 EUR
516-0577 513-3203 516-0578
Bottle top vacuum filtration systems, Nalgene®
Filters Bottle Top Filters
PS, reusable bottle top vacuum filtration funnels suitable for analytical or sterile filtration, with vacuum or pressure.
  • With graduation and screw thread
  • Removable membrane support plate
  • PP tubing adapter
513-3203: For neck size 33 mm
516-0578 and 516-0577: For neck size 45 mm
PP tubing adapter
513-3203: for neck size 33 mm
516-0578: for neck size 45 mm
Ordering information: The filter is not included and has to be ordered separately. It should have a diameter of 47 - 50 mm.