Screw caps, ventilated, for disposable erlenmeyer flasks, Nalgene®
Supplier: Thermo Fisher Scientific
4114-0038 4114-0045
214-1929EA 34.9 EUR
214-1929 214-1930
Screw caps, ventilated, for disposable erlenmeyer flasks, Nalgene®
Closures Bottle Closures
HDPE, blue with PTFE membrane, sterile.
  • Sterile packaging, each cap is packed in its own heat-sealed polybag
  • Hydrophobic 0,2 µm PTFE membrane has been heat-welded for effective sealing
  • Only suitable for use with Nalgene® sterile disposable erlenmeyer flasks
Ventilated caps ensure sterile exchange of gases for shakers and cell cultures in suspension.