Microcarrier spinner flasks, Magna-Flex®, WHEATON®
Supplier: DWK Life Sciences
356831 356830 356832 356834 356837 356839
734-3001EA 602 EUR
734-3001 734-3000 734-3002 734-3003 734-3004 734-3005
Microcarrier spinner flasks, Magna-Flex®, WHEATON®
Flasks Spinner Flasks
Borosilicate glass.
  • Two large side arms with screw cap closures allow easy sampling
  • Units 500 ml and larger have 45 mm side arms to be used as an air vent, media inlet or outlet, inoculation port, pH probe, etc
  • All flasks have been proportioned to provide a headspace of 1:1 or greater
  • Removable stainless steel pin immobilises the impeller during handling or decanting to prevent damage to cells or microcarriers
  • Size 125 to 1000 ml are graduated in 50 ml increments; sizes 3000 to 6000 ml are graduated in 500 ml increments
These flasks feature a flexMore Product Information